Learn More
Merck MultiScreen™ 96-Well Assay Plates for ELISPOT Assays
Give optimal results in immunological ELISPOT (enzyme-linked immunospot) assays by providing high protein-binding capacity with low background staining
Brand:  Merck MSIPS4W10
 View more versions of this product
Product Code. 10402362
  • 3995.00 DKK / Pack of 10
Only null left
Estimated Shipment: 20-08-2025
Description
Give optimal results in immunological ELISPOT (enzyme-linked immunospot) assays by providing high protein-binding capacity with low background staining
  • Membrane-to-plate seal ensures no reagent crosstalk and limited optical crosstalk Patented underdrain support structure allows liquid volumes to be maintained for incubation times of up to 21 days
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
Hydrophobic PVDF
0.45 μm
White
Acrylic
Sterile
ELISPOT Assays
Yes
8 x 12
MultiScreen
MultiScreen HTS
96
0.28 cm2
10/Pk.
Documents
One moment while we fetch your results.
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Merck MultiScreen™ 96-Well Assay Plates for ELISPOT Assays > Multiscreen HTS; ELISPOT; Acrylic; Immobilon-IP PVDF; 0.45μm; White; 10/Pk.