Learn More
Edwards Vacuum™ EMF10 Mist Element
For use with the EMF10 Oil Mist Exhaust Filter.
Brand:  Edwards Vacuum™ A22304198
Product Code. 10529808
  • 620.00 DKK / Each
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Description
  • Rated flow of 12 m3h
  • Maximum back pressure of 12 psig
  • Relief pressure of 20 psig
  • Replacement/spare part
  • Manufactured from epoxy impregnated borosilicate glass fiber
Specifications
Specifications
EMF10 Oil Mist Element
Epoxy-impregnated Borosilicate Glass Fiber
12 oz.
Used to separate and trap oil mist and odors discharged with the exhaust from oil sealed rotary vacuum pumps.
Back pressure of 12 psig, relief pressure of 20 psig
Product Title
Edwards Vacuum™ EMF10 Mist Element > For use with: E-LAB 2 Rotary Pump