Learn More
Memmert™ Entry Port 23 mm, Standard Position Left Side Centre/Top, For Un30Pa
Use these entry ports with convection ovens. Memmert™ Convection and Universal Oven Entry Points can be closed by flap in special positions at the back.
Brand: Memmert™ UN30paF1
This item is not returnable.
View return policy
Description
- Entry port with clear diameter
- For Memmert Ovens
Specifications
Models 30, 55, 75, 110, 160, 260, 450, 750, 1060 | |
Stainless Steel | |
23 mm |
Entry Port | |
0.9 in. |
Product Content Correction
Product Title
Memmert™ Entry Port 23 mm, Standard Position Left Side Centre/Top, For Un30Pa >
Spot an opportunity for improvement?Share a Content Correction