Learn More
Memmert Entry port 23mm, flap & stopper closure, standardposition left side centre/centre, for UN30pa
Brand:  Memmert UN30paF0
1626.49 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Entry port 23mm, flap & stopper closure, standardposition left side centre/centre, for UN30pa
Product Code. 16384397
  • 1675.00 DKK / Each
Only null left
Estimated Shipment: 17-09-2025
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Memmert Entry port 23mm, flap & stopper closure, standardposition left side centre/centre, for UN30pa >