Learn More
Memmert™ Entry Port 38 mm for UF260
Optional entry port for Memmert Ovens.
Brand:  Memmert™ UF260F7
Product Code. 17549145
  • 1630.00 DKK / Each
Estimated Shipment: 03-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Entry port with 38 mm clear diameter
  • Flap and stopper closure
  • Custom position TBC in the back
Specifications
Specifications
UF260 Oven
Entry Port
38 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Entry Port 38 mm for UF260 > For Use With: UF260