Learn More
Eppendorf™ 250mL Round Buckets for S-4-72 Rotor
For use with rotor in Eppendorf Model 5804/5810 centrifuges
Brand:  Eppendorf™ 5804747003
8480.00 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 13246589
  • 8513.92 DKK / Pack of 4
Description
  • Maximum speed of 3,234xg (4,200 rpm)
  • 250mL (8.45 oz.) capacity
  • 16cm (6.3 in.) radius
Order Info
Pack of 4
TRUSTED_SUSTAINABILITY
Specifications
Specifications
S-4-72 rotor for 5804/5810 centrifuge
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Eppendorf™ 250mL Round Buckets for S-4-72 Rotor > Round Bucket; 250mL