Learn More
Eppendorf™ Eppendorf™ Aerosol-tight Caps
Use these caps for 150mL round buckets. Eppendorf™ Aerosol-tight caps are for use with Rotor S-4-104, Rotor S-4x750 and Rotor S-4x1000.ids
Brand: Eppendorf™ 5820747005
1507.00 DKK valid until 2025-07-04
Use promo code "24147" to get your promotional price.
Description
Come in a pack of two
Specifications
Microtitor plate with 750mL bucket in S-4-104 rotor with 5810/5810R centrifuge |
Promotions
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59% -
Multiple Manufacturers
Cancer Research Campaign
Save big on essential products for cancer researchPromo Code: 24147Expires: 2025-07-31
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13