Learn More
Equilin, MedChemExpress
MedChemExpress Equilin (7-Dehydroestrone) is an important member of the large group of oestrogenic substances and is chemically related to menformon (oestrone). Equilin increases the growth of cortical neurons via an NMDA receptor-dependent mechanism.

3645.78 DKK - 5720.46 DKK
Specifications
Chemical Name or Material | Equilin |
---|---|
CAS | 474-86-2 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C18H20O2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16488656
![]() |
Medchem Express
HY-B1176 |
100 mg |
5720.46 DKK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16434567
![]() |
Medchem Express
HY-B1176 |
50 mg |
3645.78 DKK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Equilin (7-Dehydroestrone) is an important member of the large group of oestrogenic substances and is chemically related to menformon (oestrone). Equilin increases the growth of cortical neurons via an NMDA receptor-dependent mechanism.Specifications
Equilin | |
White | |
C18H20O2 | |
DMSO : 100 mg/mL (372.65 mM; Need ultrasonic) | |
268.35 | |
99.0% | |
Powder |
474-86-2 | |
Research | |
7-Dehydroestrone | |
C[C@]1([C@](CC2)([H])C3=CCC4=C(C=CC(O)=C4)[C@@]3([H])CC1)C2=O | |
268.35 | |
Research |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.