Learn More
Lenz Laborglasinstrumente™ Erlenmeyer Flasks with Conical Ground Joint
For the safe and easy mixing, pouring and storage of chemicals in the laboratory. Lenz Laborglasinstrumente™ Erlenmeyer Flasks with Conical Ground Joint are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 03024549
 View more versions of this product
Product Code. 10712522
  • 2035.00 DKK / Pack of 10
Only null left
Estimated Shipment: 01-07-2025
Description
  • Acc. to DIN EN ISO 4797
  • With scale (approx. volume/mL)
  • With inscription label
  • Corrosion-free and resistant to thermal shock
  • With conical joint, featuring high taper accuracy and excellent tightness
Specifications
Specifications
250 mL
Erlenmeyer Flask, 250mL, 85 mm dia., 45/40 NS Socket , Duran Glass, with Conical Ground Joint, Inscription Label
85 mm
Borosilicate Glass, Duran Glass
45/40 NS
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Erlenmeyer Flasks with Conical Ground Joint > Capacity: 250mL; 45/40 NS Socket