Learn More
Lenz Laborglasintrumente Erlenmeyer Flasks With Ground Joint, Amber
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 03023070
This item is not returnable.
View return policy
Description
- Acc. to DIN EN ISO 4797
- With scale (approx. volume /ml)
- With inscription label
- Material: DURAN™
Specifications
1 L | |
NS 29/32 | |
131 mm |