Learn More
Lenz Laborglasintrumente Erlenmeyer Flasks With Ground Joint, Amber
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 03023046
 View more versions of this product
Product Code. 11993371
  • 3490.00 DKK / Pack of 10
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Acc. to DIN EN ISO 4797
  • With scale (approx. volume /ml)
  • With inscription label
  • Material: DURAN™
Specifications
Specifications
200 mL
NS 29/32
79 mm