missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERMP1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
2756.95 DKK - 4398.37 DKK
Specifications
Antigen | ERMP1 |
---|---|
Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18310925
|
Bio-Techne
NBP3-17585-25UL |
25 μg |
2756.95 DKK
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18395724
|
Bio-Techne
NBP3-17585-100UL |
100 μg |
4398.37 DKK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ERMP1 Polyclonal antibody specifically detects ERMP1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
ERMP1 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
endoplasmic reticulum metallopeptidase 1, plant and worm proteins) | |
This antibody was developed against Recombinant Protein corresponding to amino acids: LGLALYLIALRTLVQLSLQQLVLRGAAGHRGEFDALQARDYLEHITSIGPRTTGSPENEILTVHYLLEQIKLIEVQSNSLHKISVDVQRPTGSFSIDFLGGFTSYYDNITNVVVKLEPRDGAQHAVLAN | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Purified | |
RUO | |
PBS, pH 7.2, 40% glycerol | |
79956 | |
IgG | |
Affinity purified |