Learn More
Estropipate, MedChemExpress
MedChemExpress Estropipate is a form of estrogen, used to treat symptoms of menopause, also used to prevent osteoporosis.
Brand:  Medchem Express HY-B1361
Encompass_Preferred
 View more versions of this product
Product Code. 16428698
  • 523.71 DKK / 10mg
Explore more special offers
Description
Description
Estropipate is a form of estrogen, used to treat symptoms of menopause, also used to prevent osteoporosis.
Specifications
Specifications
Estropipate
White
C22H32N2O5S
Piperazine estrone sulfate Estrone sulfate piperazine salt
C[C@]1([C@](CC2)([H])[C@]3([H])CCC4=C(C=CC(OS(=O)(O)=O)=C4)[C@@]3([H])CC1)C2=O.N5CCNCC5
436.56
Research
7280-37-7
Research
10 mg
DMSO : 110 mg/mL (251.97 mM; Need ultrasonic)
436.56
99.0%
Solid
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.