Learn More
DWK Life Sciences ETFE Sealing Plug 1/4 in. - 28 Thread
ETFE sealing plug for 3-port HPLC GL 45 delivery cap
Brand: DWK Life Sciences 292310003
Description
ETFE Sealing Plug 1/4 in. - 28 thread for 3-port HPLC GL 45 delivery cap
- chemically resistant
Specifications
Supply of solvents to HPLC equipment; Safe transfer of liquid media within a closed and sterile system; Chemical and thermal reactions; Storage and transport of media, APIs and reagents; Transport and shipping | |
ETFE | |
1/4 in. - 28 |