missing translation for 'onlineSavingsMsg'
Learn More
Ethanol 70% +IPA, Sterile, Ph.Eur. 1 L, Solveco Product Code.: 12624613
compliance-icons compliance-icons

Ethanol 70% +IPA, Sterile, Ph.Eur. 1 L, Solveco

Product Code. 12624613
12 x 1 L, HDPE Bottle, 12 L
Click to view available options
Quantity:
12 x 1 L
Packaging:
HDPE Bottle
Unit Size:
12 L
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Product Code. 12624613

Brand: Solveco Chemicals 1050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Solveco Sterile Ethanol D70 is a sterile-filtered, denatured ethanol solution (Ph.Eur.) with a 70% concentration, diluted with Water for Injection. The product is aseptically produced in a Laminar Airflow (LAF) unit according to current Good Manufacturing Practice (GMP) regulations.

Solveco Etanol Steril D70 is a sterile-filtered, denatured ethanol with a concentration of 70%, diluted with Water For Injection. The product is aseptically produced in a laminar air flow (LAF) environment in accordance with current Good Manufacturing Practices (GMP). It meets and is tested according to the quality requirements of the European Pharmacopoeia concerning ethanol content, density, sterility, and particle count. Solveco recommends Etanol Steril D70 for any application requiring a sterile product, such as cleaning and disinfecting clean rooms. The product is supplied with a batch-specific GMP certificate and analysis certificate. Traceability includes the raw materials used. The product can be combined with a sterile trigger spray or sterile spray nozzle to optimize disinfection results.

Specifications

CAS 64-17-5
Color Colorless
Density 0.8834–0.8884 at 20°C
Boiling Point 78°C
Quantity 12 x 1 L
Packaging HDPE Bottle
Residue after Evaporation ≤25 ppm
Molecular Formula C2H6O
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES CCO
IUPAC Name ethanol
Molecular Weight (g/mol) 46.069
PubChem CID 702
ChEBI CHEBI:16236
Formula Weight 46.07g/mol
Percent Purity 70%
Grade Sterile
Chemical Name or Material Ethyl Alcohol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Denaturerad Etanol 70% (denaturerad med Isopropanol)
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Propan-2-ol, Etanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethanol 70% +IPA, Sterile, Ph.Eur. 1 L, Solveco >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.