missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol Absolute, Solveco
Click to view available options
Quantity:
3 x 5 L
12 x 1 L
Unit Size:
Pack of 12
Pack of 3
Description
Well suited for routine laboratory and analysis. Solveco Ethanol Absolute is fermented and rectified ethanol with very high purity. This PA quality solution has a natural origin.
Specifications
Specifications
Absorbance | ≤0.500 (UV 5 cm, 240 nm) |
Color | Colorless |
Density | ≤0.790 g/cm3 |
Boiling Point | 78°C |
Quantity | 12 x 1 L |
Packaging | HDPE Bottle |
Formula Weight | 46.07g/mol |
Percent Purity | 99.9% |
Grade | AG |
Chemical Name or Material | Ethyl Alcohol |
Safety and Handling

Product Identifier
- Etanol 99,5%
- Danger
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ethanol Absolute, Solveco >
Spot an opportunity for improvement?Share a Content Correction