missing translation for 'onlineSavingsMsg'
Learn More

Ethanol Absolute, ACS Grade, Scharlab

Product Code. 12449171
Click to view available options
Quantity:
5 L
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Used in solvents, as a disinfectant, for pharmaceutical use, synthesis of organic products, and in perfumery.

  • Synonyms: Ethyl alcohol, Methylcarbinol, Spirit, Spirit of wine
  • Molecular Formula: C2H5OH
  • Molar Mass: 46.07g/mol
  • CAS: 64-17-5
  • EIN ECS No.: 200-578-6
  • Dielectric Constant: 24.3 at 25°C
  • Reducing Substances: Passes test

Reacts explosively with strong oxidizing agents including chromium trioxide, hydrogen peroxide and inorganic acids such as nitric acid. Can react violently with metals such as magnesium, sodium and potassium to produce hydrogen. May damage gaskets, painted surfaces, protective and sealing grease coatings, materials, rubber and some plastics.

Specifications

CAS 64-17-5
Quantity 5 L
Molecular Formula C2H6O
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES CCO
IUPAC Name ethanol
Molecular Weight (g/mol) 46.069
PubChem CID 702
ChEBI CHEBI:16236
Formula Weight 46.07g/mol
Show More Show Less
compliance-icons
Product Identifier
  • Ethanol absolute, ACS BASIC
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'