missing translation for 'onlineSavingsMsg'
Learn More
Ethanol, anhydrous, denatured, 85.8%, 13.3% methanol, 0.9% MIBK Product Code.: 15426165

Ethanol, anhydrous, denatured, 85.8%, 13.3% methanol, 0.9% MIBK

Product Code. 15426165
500 mL, 500mL
Click to view available options
Quantity:
500 mL
1 L
20 L
Unit Size:
1 L
20 L
500mL
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Product Code. 15426165

Brand: Thermo Scientific Alfa Aesar 036642.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

CAS: 64-17-5 | C2H6O | 46.069 g/mol

Denatured ethanol is acting as a solvent to remove the paint and ink stain on curtains, dress and upholstery. It is utilized as fuel in camping and primus stoves. It is likewise employed to clean the windows, manufacturing nail-polish and to test pectin levels in jam and jelly making.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • As a gasoline additive/substitute; octane booster
  • Very mobile liquid solvent
  • Used in lotions, tonics and perfumes
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -100°C
Color Colorless
Density 0.811g/mL
Boiling Point 80°C
Quantity 500 mL
Flash Point 11° (52°F)
Odor Like alcohol
Physical Form Liquid
Refractive Index 1.3610
Assay Percent Range 85.8%
Linear Formula CH3CH2OH
UN Number UN1993
Beilstein 1718733
Merck Index 14,3760
Solubility Information Miscible in water and most organic solvents
Formula Weight 46.08
Percent Purity 85.8%
Concentration or Composition (by Analyte or Components) 13.3% methanol, 0.9% MIBK
Chemical Name or Material Ethanol, Anhydrous, denatured
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberQ28L015 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code036642.K7,036642.AP,036642.M4,036642.K2
Lot NumberM18J001 Certificate TypeCertificate of Analysis Date25/01/2023 Product Code036636.K2,036642.K7,036636.AD,036642.AP,036636.AK
Lot NumberM13J027 Certificate TypeCertificate of Analysis Date19/01/2023 Product Code036636.K2,036642.K7,036636.AD,036642.AP,036636.AK
Lot NumberM12J043 Certificate TypeCertificate of Analysis Date18/01/2023 Product Code036636.K2,036642.K7,036636.AD,036642.AP,036636.AK
Lot NumberY17I005 Certificate TypeCertificate of Analysis Date05/01/2023 Product Code036636.K2,036642.K7,036636.AD,036642.AP,036636.AK

For Manufacturing and Laboratory Use. Not for beverage use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethanol, anhydrous, denatured, 85.8%, 13.3% methanol, 0.9% MIBK >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.