missing translation for 'onlineSavingsMsg'
Learn More
Ethanol, denatured, Spectrophotometric Grade, 90%, packaged under Argon in resealable ChemSeal™ bottles Product Code.: 15424905

Ethanol, denatured, Spectrophotometric Grade, 90%, packaged under Argon in resealable ChemSeal™ bottles

Product Code. 15424905
250 mL, 250mL
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Product Code. 15424905

Brand: Thermo Scientific Alfa Aesar 044014.AK

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

CAS: 64-17-5 | C2H6O | 46.069 g/mol

Ethanol, anhydrous, denatured is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 64-17-5, 67-56-1, 67-63-0
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point °C to 90°C
Color Colorless
Appearance Colorless liquid
Density 0.789
Boiling Point 78°C
Quantity 250 mL
Flash Point 11°C (52°F)
Ignition Point 455°C (851°F)
Odor Alcohol-like
Physical Form Liquid
Refractive Index 1.361
Assay Percent Range 90%
Vapor Pressure 57 hPa (43mm Hg) at 20°C
UN Number 1987
Beilstein 1718733
Solubility Information Miscible with water and most organic solvents.
Formula Weight 46.08
Grade Spectrophotometric
Concentration or Composition (by Analyte or Components) Ethyl alcohol: 90.0%; Methanol: 5.0%; 2-Propanol: 5.0%
Chemical Name or Material Ethanol, denatured
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberS05L722 Certificate TypeCertificate of Analysis Date16/06/2025 Product Code044014.K2,044014.AK
Lot NumberJ30W962 Certificate TypeCertificate of Analysis Date18/09/2024 Product Code044014.K2,044014.AK
Lot NumberJ15R784 Certificate TypeCertificate of Analysis Date16/10/2023 Product Code044014.K2,044014.AK
Lot NumberP30H763 Certificate TypeCertificate of Analysis Date09/05/2023 Product Code044014.K2,044014.AK
Lot NumberW20F745 Certificate TypeCertificate of Analysis Date11/11/2022 Product Code044014.K2,044014.AK

For Manufacturing and Laboratory Use. Not for beverage use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethanol, denatured, Spectrophotometric Grade, 90%, packaged under Argon in resealable ChemSeal™ bottles >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.