missing translation for 'onlineSavingsMsg'
Learn More

Ethanol (Untaxed), Purum, absolute ethanol, denaturated with 1% toluene, A15 T1, ≥99.8% (based on denaturant-free substance), for license holders only, Honeywell

Product Code. 15690630
Click to view available options
Quantity:
1 L
2.5 L
10 L
4 x 2.5 L
Packaging:
Glass Bottle
Poly Pail
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Chemical Identifiers

CAS 64-17-5, 108-88-3
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

CAS Min % ≥50.0000%
CAS Max % ≤100.0000%, <3.0000%
Density 0.80 g/cm3
Boiling Point 78°C
Quantity 10 L
Flash Point 9°C
Packaging Poly Pail
Linear Formula CH3CH2OH
UN Number UN1170
Beilstein 1718733
Water max. 0.3%
Formula Weight 46
Percent Purity >99.8%
Grade purum
Assay Gew.
Chemical Name or Material Ethanol (Untaxed), Purum, absolute ethanol, denaturated with 1% toluene, A15 T1, ≥99.8% (based on denaturant-free substance), for license holders only
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.