missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol (Untaxed), Purum, fine spirit, denaturated with 4.8% isopropanol, F25 IPA1, ∼96% (based on denaturant-free substance), for license holders only, Honeywell
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Specifications
Specifications
CAS | 64-17-5, 67-63-0 |
CAS Min % | ≥50.0000% |
CAS Max % | ≤100.0000%, <10.0000% |
Density | 0.79 - 0.81 g/cm3 |
Boiling Point | 77°C to 79°C |
Quantity | 1 L |
Flash Point | 14°C |
Packaging | Glass Bottle |
Molecular Formula | C2H6O |
MDL Number | MFCD00011324 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction