missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Ethidium Bromide Solution (10mg/mL)
Ethidium Bromide Solution (10mg/mL), Molecular biology grade
486.00 DKK - 1440.00 DKK
Specifications
CAS | 1239-45-8 |
---|---|
Detection | 15-30 min with 0.5mg/mL solution |
Color | Dark Red/Purple |
Concentration | 10 mg/mL |
Product Type | Fluorescent DNA/RNA Stain |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11417060
|
MP Biomedicals™
11ETBC1001 |
10 mL |
486.00 DKK
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11427060
|
MP Biomedicals™
11ETBC1002 |
6 x 10 mL |
1440.00 DKK
60mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Concentration : 0.95 – 1.05%
- Purity (on dry basis) : > 98.0%
- RNase, DNase, Protease free
- Fluorescent DNA and RNA intercalating agent.
- Used as a dye for DNA and RNA electrophoresis.
- Storage: Store at + 4°C protected from light.
- A 10 mg/mL Ready-To-Use solution of EtBr in specially filtered, deionized water.
- Ethidium bromide has been used in a number of fluorimetric assays for nucleic acids.
- It has been shown to bind to single-stranded DNA (although not as strongly) and triple-stranded D
- Ethidium bromide (EtBr) is useful for all applications requiring nucleic acid staining including CsCl plasmid DNA isolation and is the most commonly used nucleic acid stain for PAGE or agarose gel electrophoresis.
Specifications
1239-45-8 | |
Dark Red/Purple | |
Fluorescent DNA/RNA Stain | |
Nucleic Acid Electrophoresis; Blotting & Detection; Nucleic Acidgel Electrophoresis & Blotting | |
Store at +4°C, protect from light. |
15-30 min with 0.5mg/mL solution | |
10 mg/mL | |
Liquid | |
394.3 |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Acute toxicity Category 3
- Germ cell mutagenicity Category 2
Hazard Statement
- H331-Toxic if inhaled.
- H341-Suspected of causing genetic defects.
Precautionary Statement
- P102-Keep out of reach of children.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethidium bromide
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ Ethidium Bromide Solution (10mg/mL)