missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 4,4,4-trifluoro-2-butynoate, 97% Product Code.: 15420228

Ethyl 4,4,4-trifluoro-2-butynoate, 97%

Product Code. 15420228
5 g, 5g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15420228

Brand: Thermo Scientific Alfa Aesar H53425.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 79424-03-6 | C6H5F3O2 | 166.099 g/mol

Ethyl 4,4,4-trifluoro-2-butynoate is the suitable reagent used to investigate the regioselectivity of the insertion reaction with cyclometalated iridium and rhodium complexes. It may be used in the preparation of ethyl (Z)-3-iodo-4,4,4-trifluoro-2-butenoate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4,4,4-trifluoro-2-butynoate is the suitable reagent used to investigate the regioselectivity of the insertion reaction with cyclometalated iridium and rhodium complexes. It may be used in the preparation of ethyl (Z)-3-iodo-4,4,4-trifluoro-2-butenoate.

Solubility
Soluble in water (partly).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79424-03-6
Molecular Formula C6H5F3O2
Molecular Weight (g/mol) 166.099
MDL Number MFCD00192167
InChI Key SFDRHPQGYUYYNX-UHFFFAOYSA-N
PubChem CID 3597254
IUPAC Name ethyl 4,4,4-trifluorobut-2-ynoate
SMILES CCOC(=O)C#CC(F)(F)F

Specifications

Density 1.162
Boiling Point 96°C to 98°C
Flash Point 6°C (43°F)
Linear Formula CF3C≡CCO2CH2CH3
Refractive Index 1.35
Quantity 5 g
UN Number UN3272
Beilstein 3539414
Synonym ethyl 4,4,4-trifluoro-2-butynoate, 4,4,4-trifluoro-2-butynoic acid ethyl ester, 4,4,4-trifluoro-but-2-ynoic acid ethyl ester, pubchem10880, acmc-1bc5r, ethyl4,4,4-trifluoro-2-butynoate, ethyl 4,4,4-trifluoro-2-butynate, trifluoro-2-butynoic acid ethyl ester, ethyl 4,4,4-trifluorobutynoate, ethyl 4,4,4-tris fluoranyl but-2-ynoate
Solubility Information Soluble in water (partly).
Formula Weight 166.1
Percent Purity 97%
Chemical Name or Material Ethyl 4,4,4-trifluoro-2-butynoate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberX02F032 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH53425.03,H53425.06
Lot NumberW25B013 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH53425.03,H53425.06
Lot NumberT11A017 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH53425.03,H53425.06
Lot NumberX25C039 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH53425.03,H53425.06
Lot NumberK03X014 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH53425.03,H53425.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 4,4,4-trifluoro-2-butynoate, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.