missing translation for 'onlineSavingsMsg'
Learn More
Ethyl 5-methylisoxazole-3-carboxylate, 95%, Thermo Scientific™ Product Code.: 11480743

Ethyl 5-methylisoxazole-3-carboxylate, 95%, Thermo Scientific™

Product Code. 11480743
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11480743

Brand: Thermo Scientific Alfa Aesar A13226.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ethyl 5-methylisoxazole-3-carboxylate, It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3209-72-1
Molecular Formula C7H9NO3
Molecular Weight (g/mol) 155.15
MDL Number MFCD00085080
InChI Key OCCIGHIQVMLYBZ-UHFFFAOYSA-N
Synonym ethyl 5-methylisoxazole-3-carboxylate, 3-isoxazolecarboxylic acid, 5-methyl-, ethyl ester, 5-methyl-isoxazole-3-carboxylic acid ethyl ester, 5-methylisoxazole-3-carboxylate ethyl, vitas-bb tbb000645, pubchem8773, 5-methylisoxazole-3-carboxylicacidethylester, acmc-1ctp0, ksc226e3b, 3-ethoxycarbonyl-5-methylisoxazole
PubChem CID 76677
IUPAC Name ethyl 5-methyl-1,2-oxazole-3-carboxylate
SMILES CCOC(=O)C1=NOC(C)=C1

Specifications

Melting Point 27°C to 31°C
Boiling Point 77°C to 79°C (0.5 mmHg)
Flash Point 109°C (228°F)
Quantity 1 g
Solubility Information It is insoluble in water.
Formula Weight 155.15
Percent Purity 95%
Chemical Name or Material Ethyl 5-methylisoxazole-3-carboxylate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10192705 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethyl 5-methylisoxazole-3-carboxylate, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.