missing translation for 'onlineSavingsMsg'
Learn More
ethyle6-chloroimidazo[1,2-b]pyridazine-2-carboxylate, 97%, Thermo Scientific™ Product Code.: 10455512

ethyle6-chloroimidazo[1,2-b]pyridazine-2-carboxylate, 97%, Thermo Scientific™

Product Code. 10455512
250 mg, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10455512

Brand: Thermo Scientific Maybridge MO00216CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 64067-99-8
Molecular Formula C9H8ClN3O2
Molecular Weight (g/mol) 225.632
MDL Number MFCD03086230
InChI Key AZEPEWATPYRPBM-UHFFFAOYSA-N
Synonym ethyl 6-chloroimidazo 1,2-b pyridazine-2-carboxylate, imidazo 1,2-b pyridazine-2-carboxylic acid, 6-chloro-, ethyl ester, 6-chloroimidazo 1,2-b pyridazine-2-carboxylic acid ethyl ester, 6-chloro-imidazo 1,2-b pyridazine-2-carboxylic acid ethyl ester, pubchem19273, acmc-209nk3, ethyl 6-chloroimidazo 1,2-b pyridazin-2-carboxylate, ethyl 6-chloroimidazo 1,2-b pyridazin-2-ylcarboxylate, ethyl 6-chloro-4-hydroimidazo 1,2-e pyridazine-2-carboxylate, ethyl 6-chloroimidazo 2,1-f pyridazine-2-carboxylateShow MoreShow Less
PubChem CID 2779714
IUPAC Name ethyl 6-chloroimidazo[1,2-b]pyridazine-2-carboxylate
SMILES CCOC(=O)C1=CN2C(=N1)C=CC(=N2)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 225.63
Percent Purity 97%
Chemical Name or Material Ethyl 6-chloroimidazo[1, 2-b]pyridazine-2-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
ethyle6-chloroimidazo[1,2-b]pyridazine-2-carboxylate, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.