missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic acid disodium zinc salt tetrahydrate, Honeywell™

Product Code. 15664280
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

for complexometry,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 176736-49-5
Quantity 100 g
Molecular Formula C10H20N2Na2O12Zn
Linear Formula (NaOOCCH2)2NCH2CH2N(CH2COO)2Zn · 4H2O
MDL Number MFCD00151728
UN Number NONH for all modes of transport
Synonym disodium zinc ethylenediaminetetraacetate tetrahydrate, edta-na2zn, ethylenediaminetetraacetic acid disodiu&, zinc 2+ disodium ion 4-edta tetrahydrate, ethylenediaminetetraacetic acid disodium zinc salt tetrahydrate
InChI Key ZEVLVCFKKCNROJ-UHFFFAOYSA-J
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].O.O.O.O.[Na+].[Na+].[Zn+2]
IUPAC Name disodium;zinc;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;tetrahydrate
Molecular Weight (g/mol) 471.632
PubChem CID 53393534
Formula Weight 471.64g/mol
Percent Purity ≥98%
Chemical Name or Material Ethylenediaminetetraacetic acid disodium zinc salt tetrahydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.