Learn More
Fisherbrand™ Sealing Mat for 96-Well Deep Square Nonsterile Microplates
Fisherbrand™ Sealing Mat for 96-Well Deep Square Nonsterile Microplates is a reusable EVA sealing cap that fits onto the top of the plate to retain the sample in the well during transit or to stop the well and its contents from becoming contaminated.
Brand:  Fisherbrand™ 219004
Encompass
Product Code. 11521963
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • For friction sealing of plates during transport or storage
  • Cap mats are an economic alternative to thermal or adhesive seals
  • Cap mats can be sterilized and reused
  • Constructed of high quality ethylene-vinyl acetate (EVA)
  • Rnase/Dnase free
Specifications
Specifications
White
Non-sterile
50
Square
EVA
96
96 Well Square Sealing Cap
96-well Square Deepwell Plate
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Sealing Mat for 96-Well Deep Square Nonsterile Microplates