Learn More
EVOS™ 50X Oil Objective, achromat, coverslip-corrected
For general applications and image with different magnification
Brand: EVOS™ AMPFOP050
This item is not returnable.
View return policy
Description
- Coverslip-corrected objective that is optimized for imaging through #1.5 coverslips
- Offers optical performance from visible light to near infrared light
- Meets needs across spectrum of magnifications and optical specifications
Specifications
Objective Lens | |
EVOS™ FL Color Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System | |
0.19 mm | |
50X | |
1 Ea. |
EVOS | |
Vessels, Slides, Cell Culture Dishes, Flasks, Microtiter Well Plates | |
Plan Achromat | |
0.95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction