Learn More
Invitrogen™ EVOS™ Light Cube, CFP-YFP EM 2.0
EVOS LED light cubes provide superior illumination uniformity across the field of view, resulting in exceptional tile-stitch image quality. At the same time, optimized spectral fidelity across channels significantly improves data quality by minimizing undesired bleed-through.
Brand: Invitrogen™ AMEP4969
Description
- Second-generation EVOS LED light cubes provide superior illumination uniformity across the field of view, resulting in exceptional tile-stitch image quality
- At the same time, optimized spectral fidelity across channels significantly improves data quality by minimizing undesired bleed-through
- Like the original design, they combine condensing optics and hard-coated filters into one light engine that delivers digitally controlled fluorophore excitation
- These LED light cubes are backwards compatible with older EVOS and Countess systems. and can be used with first-generation light cubes
- Like the original light cubes, these LED cubes are user-changeable and automatically recognized by the instrument
- The EVOS Light Cube, CFP-YFP EM 2.0, is ideal for use with CFP-YFP FRET emission, and other fluorophores with excitation and emission maxima near 445 and 542 nm
- The LED bulbs on the EVOS systems are rated for >50,000 hours, compared to 300 hours for a typical mercury bulb and 1,500 hours for a metal halide bulb
EVOS LED light cubes offer these important advantages:
- Uniform illumination across the entire field of view for superb image quality
- Optimized spectral compatibility to maximize true signal detection, while minimizing bleed-through for the best possible data quality
- A short light path that affords superior detection of fluorescent signals
- Stable illumination intensity over 50,000 hours that helps experimental reproducibility
Specifications
LED Light Cube | |
Ex: 445/45 nm Em: 542/27 | |
Condensing Optics, Hard-coated Filters |
EVOS | |
Countess™ III FL Automated Cell Counter, EVOS™ M5000 Imaging System, EVOS™ M7000 Imaging System | |
1 each |
Product Suggestions
Customers who viewed this item also viewed