Learn More
Memmert™ Extended Overtemperature Protection for SN30
Extended overtemperature protection with integrated Pt100 sensor for independent temperature monitoring.
Brand:  Memmert™ SN30A6
Product Code. 17548575
  • 1425.00 DKK / Each
Estimated Shipment: 07-04-2025
to see stock.
Explore more special offers
Description
  • Accessory
  • With integrated Pt100 sensor
  • For models with SingleDISPLAY
Specifications
Specifications
SN30 Sterilizer
Integrated Pt100 Sensor