Learn More
Memmert™ Extended Overtemperature Protection for UF55
Extended overtemperature protection with integrated Pt100 sensor for independent temperature monitoring.
Brand: Memmert™ UF55A6
1387.74 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Accessory
- With integrated Pt100 sensor
- For models with SingleDISPLAY
Specifications
UF55 Oven | |
Integrated Pt100 Sensor |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Extended Overtemperature Protection for UF55 > For Use With: UF55
Spot an opportunity for improvement?Share a Content Correction