Learn More
Memmert™ Extension of DAkkS Factory Calibration for UF260
Extension of DAkkS factory calibration by an additional freely selectable temperature for Memmert Ovens.
Brand: Memmert™ UF260E39697
This item is not returnable.
View return policy
Description
- Extension of DAkkS calibration at Memmert factory by an additional freely selectable temperature value according to method C (DKD-R 5-7)
Specifications
UF260 Oven | |
Factory Calibration Extension |
Product Content Correction
Product Title
Memmert™ Extension of DAkkS Factory Calibration for UF260 > For Use With: UF260
Spot an opportunity for improvement?Share a Content Correction