Learn More
Lenz Laborglasintrumente Extractor Heads Acc. to Thielepape
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
2010.00 DKK - 2205.00 DKK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
10040241
|
Lenz Laborglasintrumente
05405143 |
150 mL |
2010.00 DKK
Each |
Estimated Shipment: 10-06-2025
Log in to see stock. |
||||||
10274241
|
Lenz Laborglasintrumente
05405149 |
250 mL |
2205.00 DKK
Each |
Estimated Shipment: 10-06-2025
Log in to see stock. |
||||||
Description
- Extractor heads acc. to Thielepape
- With NS 2-way stopcock
- Made of DURAN™ tubing
Specifications
Borosilicate Glass 3.3 | |
NS 45/40 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Lenz Laborglasintrumente Extractor Heads Acc. to Thielepape