Learn More
Lenz Laborglasintrumente Extractor Heads for Specific Heavy Solvents
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05433149
 View more versions of this product
Product Code. 10151471
  • 2400.00 DKK / Each
Estimated Shipment: 16-06-2025
Explore more special offers
Description
  • For the extraction of liquids with specific heavy solvents
  • With insert filter (porosity: 0)
  • Made of DURAN™ tubing
Specifications
Specifications
250 mL
Fritte P0
NS 45/40
Documents
One moment while we fetch your results.
Product Title
Lenz Laborglasintrumente Extractor Heads for Specific Heavy Solvents > 250 mL