Learn More
Medline Scientific™ Eyepiece for Varizoom Stereo Microscopes
With 20x magnification.
Brand:  Medline Scientific™ 7700.2004
Product Code. 12633035
  • 3035.00 DKK / Each
Estimated Shipment: 24-03-2025
to see stock.
Explore more special offers
Description
  • 20x/13mm wide-field
  • For use with Varizoom Stereo Microscopes
Specifications
Specifications
Eyepiece
Varizoom Stereo Microscopes
1 Ea.
13 mm
20X