Learn More
BINDER™ Factory Calibration Certificate
For Use With: BINDER units series C and CB
Brand: BINDER™ DL30-0402
This item is not returnable.
View return policy
Description
- O2 measurement with analyzed test gas at 5% or at specified value
- For use with C and CB series incubators
Specifications
Calibration O2 - including certificate, O2 measurement with analyzed test gas at 5 % or at specified value | |
BINDER Chambers Series CB-S, CB, CBF | |
Calibration Certificate | |
Measurement of Carbon Dioxide |
Product Content Correction
Product Title
BINDER™ Factory Calibration Certificate > For Use With: C and CB series
Spot an opportunity for improvement?Share a Content Correction