missing translation for 'onlineSavingsMsg'
Learn More
Fadraciclib, MedChemExpress Product Code.: 16460739

Fadraciclib, MedChemExpress

Product Code. 16460739
1 mg, 1mg
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16460739

Brand: Medchem Express HY101212

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fadraciclib (CYC065) is a second-generation, orally available ATP-competitive inhibitor of CDK2/CDK9 kinases with IC50s of 5 and 26 nM, respectively.

Fadraciclib (CYC065) is a second-generation, orally available ATP-competitive inhibitor of CDK2/CDK9 kinases with IC50s of 5 and 26 nM, respectively.

Specifications

Chemical Name or Material Fadraciclib
CAS 1070790-89-4
Color White
Purity Grade Notes Research
Molecular Formula C21H31N7O
Quantity 1 mg
Synonym CYC065
Solubility Information DMSO : ≥ 100 mg/mL (251.56 mM)
SMILES C[C@@H](O)[C@@H](NC1=NC(NCC2=C(C)C=C(C)N=C2)=C3N=CN(C(C)C)C3=N1)CC
Molecular Weight (g/mol) 397.52
Formula Weight 397.52
Percent Purity 99.56%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Fadraciclib, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.