missing translation for 'onlineSavingsMsg'
Learn More

FAM102A Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

3427.56 DKK

Specifications

Antigen FAM102A
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18310846
View Documents Promotion Details
Bio-Techne
NBP3-10723-100UL
100 μg
3427.56 DKK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FAM102A Polyclonal specifically detects FAM102A in Human samples. It is validated for Western Blot.
Specifications

Specifications

FAM102A
Western Blot
Unconjugated
Rabbit
Human
bA203J24.7, C230093N12Rik, C9orf132, Early estrogen-induced gene 1 protein, Eeig1, EEIG1chromosome 9 open reading frame 132, family with sequence similarity 102, member A, MGC50853
The immunogen for Anti-FAM102A antibody is: synthetic peptide directed towards the Middle region of Human F102A. Peptide sequence SLSDLTHRRNTSTSSSASGGLGMTVEGPEGSEREHRPPEKPPRPPRPLHL
Affinity purified
Western Blot 1.0 ug/ml
Polyclonal
Purified
RUO
PBS buffer, 2% sucrose
399665
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications