missing translation for 'onlineSavingsMsg'
Learn More

FBXO24 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

1485.92 DKK - 3498.83 DKK

Specifications

Antigen FBXO24
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18302064
View Documents Promotion Details
Bio-Techne
NBP3-04452-20UL
20 μL
1485.92 DKK
20µL
Please to purchase this item. Need a web account? Register with us today!
18310507
View Documents Promotion Details
Novus Biologicals
NBP3-04452-100UL
100 μL
3498.83 DKK
100µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FBXO24 Polyclonal specifically detects FBXO24 in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

FBXO24
Polyclonal
Purified
RUO
PBS with 50% glycerol, pH7.3.
26261
IgG
Affinity purified
Western Blot
Unconjugated
Rabbit
Human, Mouse
DKFZp434I1122, F-box protein 24, F-box protein Fbx24, FBX24F-box only protein 24
Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human FBXO24 (NP_277041.1).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications