missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ FCER1A (Human) Recombinant Protein

Human FCER1A full-length ORF ( AAH05912.1, 1 a.a. - 257 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00002205-P01.25ug

 View more versions of this product

Product Code. 16134881

  • 4040.00 DKK / 25µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Sequence: MAPAMESPTLLCVALLFFAPDGVLAVPQKPKVSLNPPWNRIFKGENVTLTCNGNNFFEVSSTKWFHNGSLSEETNSSLNIVNAKFEDSGEYKCQHQQVNESEPVYLEVFSDWLLLQASAEVVMEGQPLFLRCHGWRNWDVYKVIYYKDGEALKYWYENHNISITNATVEDSGTYYCTGKVWQLDYESEPLNITVIKAPREKYWLQFFIPLLVVILFAVDTGLFISTQQQVTFLLKIKRTRKGFRLLNPHPKPNPKNN
Specifications

Specifications

AAH05912.1
Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FCER1A
GST
2205
Wheat germ expression system
25 μg
FCE1A, FcERI
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers