Learn More
Thermo Scientific™ EpiJET DNA Methylation Analysis Kit (MspI/HpaII)
Perform fast and efficient analysis of the DNA methylation status at a specific locus with the Thermo Scientific™ EpiJET™ DNA Methylation Analysis Kit (MspI/HpaII).
Brand:  Thermo Scientific™ K1441
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11388851
  • 1162.00 DKK / 200 reactions
Explore more special offers
Description
Description
5-Methylcytosine is a prominent epigenetic DNA modification which plays an important role in regulation of gene expression.
EpiJET DNA Methylation Analysis Kit (MspI/HpaII) uses the MspI and HpaII restriction enzymes to analyze DNA methylation status at a specific locus. Epi MspI and Epi HpaII are isoschizomers with differing sensitivities to CpG methylation. When the internal CpG in the 5'-CCGG-3' tetranucleotide sequence is methylated, cleavage with Epi HpaII is blocked, but cleavage with Epi MspI is not affected.
The Epi MspI and Epi HpaII enzymes are specially formulated for epigenetics studies to complete genomic DNA digestion in 1 hour.
Highlights:
Fast—complete digestion of genomic DNA in 1 hour at 37°C
Efficient—specially formulated enzymes for epigenetic studies of DNA methylation status
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10X Epi Buffer
200 reactions
DNA Methylation Analysis Kit
EpiJET™
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2809201 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK1441
Lot Number2797769 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK1441
Lot Number2793444 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK1441
Lot Number2809209 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK1441
Lot Number2828798 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK1441
Product Title
Thermo Scientific™ EpiJET DNA Methylation Analysis Kit (MspI/HpaII) > For 200 react.