Learn More
Thermo Scientific™ NoLimits 1500 bp DNA Fragment
GREENER_CHOICE
Optimize applications where a custom DNA size standard is required with NoLimits, a collection of 36 chromatography-purified individual DNA fragments ranging from 10 -20,000bp.
Brand:  Thermo Scientific™ SM1691
Product Code. 10151270
  • 692.00 DKK / 10µg
Explore more special offers
This item is not returnable. View return policy
Description
Description
NoLimits is a collection of 36 highly-purified individual DNA fragments ranging from 10 to 20,000 bp. Unlike other vendors, where custom standards are produced by PCR, Thermo Scientific NoLimits DNA fragments are produced using specifically designed plasmids purified by patented technology.
Plasmid DNA is digested with Thermo Scientific Restriction Enzymes, and the individual DNA fragments are chromatography-purified from the digestion mixture. As a result, we offer exceptionally pure DNA fragments that are free of any truncated or degraded molecules. Our custom DNA fragments are the best choice for applications, ranging from electrophoresis (both gel and capillary) to HPLC and beyond.
Highlights
  • Chromatography-purified individual DNA fragments
  • Broad range: 10 to 20,000 bp
  • Sharp peaks during capillary electrophoresis, sharp bands during gel electrophoresis
  • Precise DNA concentration 
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Agarose
No
Agarose Gels
NoLimits
DNA Fragment
10 to 20000 bp
Sustainable packaging
10 μg
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2793163 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberSM1691
Lot Number2834072 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberSM1691
Lot Number2876100 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberSM1691
Lot Number2876116 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberSM1691
Lot Number3019443 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberSM1691
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ NoLimits 1500 bp DNA Fragment > 10μg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
For Research Use Only. Not for use in diagnostic procedures.