missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferritin Rabbit anti-Human, Mouse, Rat, Clone: S03-3D9, Novus Biologicals™
Rabbit Monoclonal Antibody
2180.29 DKK - 4407.01 DKK
Specifications
Antigen | Ferritin |
---|---|
Clone | S03-3D9 |
Applications | Western Blot, ELISA, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18310495
|
Novus Biologicals
NBP3-15028-25UL |
25 μg |
2180.29 DKK
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18364352
|
Novus Biologicals
NBP3-15028-100UL |
100 μg |
4407.01 DKK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ferritin Monoclonal antibody specifically detects Ferritin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
Ferritin | |
Western Blot, ELISA, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cancer, Cellular Markers, Lipid and Metabolism | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
2495 | |
IgG | |
Affinity-purified |
S03-3D9 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
apoferritin, Cell proliferation-inducing gene 15 protein, Ferritin H subunit, ferritin heavy chain, ferritin, heavy polypeptide 1, FHC, FTHEC 1.16.3.1, FTHL6MGC104426, PIG15, placenta immunoregulatory factor, PLIF, proliferation-inducing protein 15 | |
A synthetic peptide of human Ferritin (Uniprot#: P02794) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |