missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGD2 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-99377AF750
This item is not returnable.
View return policy
Description
FGD2 Polyclonal specifically detects FGD2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FGD2 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
FGD1 family, member 2, FLJ00048, FLJ00276 protein, FLJ40929, FYVE, RhoGEF and PH domain containing 2, FYVE, RhoGEF and PH domain-containing protein 2, ZFYVE4MGC71330, Zinc finger FYVE domain-containing protein 4 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human FGD2. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
221472 | |
Store at 4°C in the dark. | |
IgG |