missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibrinogen beta chain Mouse anti-Human, Clone: 1F8E4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66186-1-IG-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is the beta component of fibrinogen, a blood-borne glycoprotein comprised of three pairs of nonidentical polypeptide chains. Following vascular injury, fibrinogen is cleaved by thrombin to form fibrin which is the most abundant component of blood clots. In addition, various cleavage products of fibrinogen and fibrin regulate cell adhesion and spreading, display vasoconstrictor and chemotactic activities, and are mitogens for several cell types. Mutations in this gene lead to several disorders, including afibrinogenemia, dysfibrinogenemia, hypodysfibrinogenemia and thrombotic tendency.Specifications
Fibrinogen beta chain | |
Monoclonal | |
1.45 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P02675 | |
FGB | |
Fibrinogen beta chain Fusion Protein Ag10205 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Western Blot | |
1F8E4 | |
Unconjugated | |
FGB | |
FGB, fibrinogen beta chain | |
Mouse | |
Protein A | |
RUO | |
2244 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fibrinogen beta chain Mouse anti-Human, Clone: 1F8E4, Proteintech > 150 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido