missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Fibroblast Growth Factor 7, human
944.00 DKK - 1654.36 DKK
Specifications
Molecular Weight (g/mol) | 18.9 kd |
---|---|
Packaging | Plastic bottle |
Form | Lyophilized |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15448609
|
Thermo Scientific Chemicals
J67052.&E |
2μg | N/A | ||||||
15438609
|
Thermo Scientific Chemicals
J67052.!E |
1654.36 DKK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
18.9 kd | |
Plastic bottle | |
Lyophilized |
RUO – Research Use Only