missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibronectin Antibody (A17), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-23524CL1
This item is not returnable.
View return policy
Description
Fibronectin Monoclonal antibody specifically detects Fibronectin in Bovine,Chicken,Human samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen)Specifications
Fibronectin | |
Monoclonal | |
CoraFluor 1 | |
CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF | |
Lysed bovine corneal endothelial cells and extracellular matrix | |
0.1 mL | |
Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
2335 | |
Store at 4°C in the dark. Do not freeze. | |
IgG1 |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
A17 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Bovine, Chicken, Human | |
Purified |