Learn More
Thermo Scientific™ Finnpipette™ Dispenser
Dose liquids from reagent bottles with dispensers that offer superior chemical and thermal resistance, and an ergonomic design that prevents work fatigue.
3330.00 DKK - 4725.00 DKK
Specifications
Product Type | Analog Bottletop Dispenser |
---|---|
Certifications/Compliance | Individual calibration certificate with serial number |
Channels | 1 |
Product Code | Brand | Precision | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Precision | Description | Price | Quantity & Availability | |||||
11448409
![]() |
Thermo Scientific™
4421120 |
2µL | Range: 0.2-1.0mL, ±6.0µL |
3330.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
12811171
![]() |
Thermo Scientific™
4421130 |
4 µL | Range: 0.4-2.0mL, ±12.0µL |
3330.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
11468409
|
Thermo Scientific™
4421140 |
10 µL | Range: 1-5mL, ±30.0µL |
3330.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
11478409
|
Thermo Scientific™
4421150 |
20µL | Range: 2-10mL, ±60.0µL |
3330.00 DKK
Each |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
11488409
|
Thermo Scientific™
4421160 |
60 µL | Range: 5-30mL, ±180µL |
4435.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
12326532
|
Thermo Scientific™
4421170 |
120µL | Range: 10-60mL, ±360µL |
4725.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
Description
Dose liquids from reagent bottles with Thermo Scientific™ Finnpipette™ Dispensers.
Features:
- Excellent chemical and thermal resistance
- Fully autoclavable†
- Exact self-locking system in the volume setting
- Dispensing head cap prevents dripping
- Ergonomic design prevents work fatigue
- Threaded suction hose for bubble-free suction
Note:
Limitations include HF, liquids that attack Halar, FEP or Hastelloy suspensions because solid particles might block the valves.
Warranty: 1 year
Specifications
Analog Bottletop Dispenser | |
Individual calibration certificate with serial number | |
1 |
Note: Not for use on hot plates.