Learn More
Thermo Scientific™ Finnpipette™ F2 Fixed Volume Single-Channel Pipettes
GREENER_CHOICE
Use these fixed volume single-channel pipettes for highly demanding applications where robustness and durability are essential.
Brand:  Thermo Scientific™ 4652130
1008.59 DKK valid until 2025-06-30
Use promo code "25076" to get your promotional price.
 View more versions of this product
Product Code. 12356552
  • 1635.00 DKK / Each
In Stock
to see stock.
Explore more special offers
Description
Use Thermo Scientific™ Finnpipette™ F2 Fixed Volume Single-Channel Pipettes for highly demanding applications where robustness and durability are essential. With a sleek, rugged design, these pipettes are built with tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light. Fully autoclavable.
For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.
Finnpipette F2: Proven reliable for long-term intensive use
Rugged and Durable
Finnpipette F2 contains tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light. Its rugged design withstands physical use without damage.
Fully Autoclavable
Decontamination of the Finnpipette F2 is simple as there is no need to disassemble the F2 for autoclaving, minimizing disruption and downtime.
Maintenance is Easy
Simply detach the tip cone for efficient daily maintenance or decontamination when not using an autoclave.
Super Blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50μL models and below.
Wide Selection of Finntip Pipette Tips
Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.
5 Year Warranty
With convenient online registration, and may vary by country.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Finntip 250, Filter 20, Flex 200, Flex Filter 30
Autoclavable
F2
Fixed Volume Pipette
FinnpipetteTM F2 fixed volume
PVDF
1
20 μL
SDS
Documents Certificates
View accessory products
Special Offers
Special Offers
  • Thermo Scientific ClipTip , Thermo Scientific ART , Thermo Scientific Finnpipette , Thermo Scientific Matrix
    Unlock Precision: Top Pipettes and Tips on Promotion!
    Promo Code: 25076
    Expires2025-06-30
    Days
    65
    Hours
    14
    Minutes
    43
    Seconds
    39
Product Content Correction
Product Title
Thermo Scientific™ Finnpipette™ F2 Fixed Volume Single-Channel Pipettes >