Learn More
3M™ First Base™ + Bump Cap
First Base™ + Bump Cap designed to provide limited head protection from bumps and minor lacerations from stationary objects. Patented flexible shell design manufactured from durable ABS plastic provides industrial bump protection to EN812.
2497.28 DKK - 3465.00 DKK
Products 10
Product Code Brand Product Type Color Price Quantity & Availability  
Product Code Brand Product Type Color Price Quantity & Availability  
17207690
View Documents
3M™
7100206581
Micro Peak Navy Blue
3465.00 DKK
Pack of 20
Estimated Shipment: 05-08-2025
Add to basket
17217690
View Documents
3M™
7100206582
Reduced Peak This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17227690
View Documents
3M™
7100206586
Standard Peak Black
3465.00 DKK
Pack of 20
Estimated Shipment: 05-08-2025
Add to basket
16863553
View Documents
3M™
7100206593
Standard Peak Royal Blue
3340.00 DKK
Pack of 20
Estimated Shipment: 05-08-2025
Add to basket
16873553
View Documents
3M™
7100207096
Micro Peak Gray
3340.00 DKK
Pack of 20
Estimated Shipment: 05-08-2025
Add to basket
17267680
View Documents
3M™
7100206559
3340.00 DKK
Pack of 20
Estimated Shipment: 05-08-2025
Add to basket
17267690
View Documents
3M™
7100208068
Reduced Peak Royal Blue
3465.00 DKK
Pack of 20
Estimated Shipment: 05-08-2025
Add to basket
17277680
View Documents
3M™
7100206560
3465.00 DKK
Pack of 20
Estimated Shipment: 05-08-2025
Add to basket
17287680
View Documents
3M™
7100206561
3340.00 DKK
Pack of 20
Estimated Shipment: 05-08-2025
Add to basket
17297680
View Documents
3M™
7100206562
Reduced Peak Black
3465.00 DKK
Pack of 20
Estimated Shipment: 05-08-2025
Add to basket
Description
Description
  • Provides industrial bump protection to EN812.
  • Flexible shell made with ABS plastic, can be removed for cleaning.
  • Airflow channels for ventilation.
  • Absorbent terry toweling sweatband.
  • Available in different peak lengths for a variety of applications.
  • Adjustable elastic strap for a personal fit.
  • Optional logo customization areas.
  • Available in different peak lengths for a variety of applications.
  • Adjustable elastic strap for a personal fit.
  • Belt clip accessory available.
Documents
Product Certifications
Product Content Correction
Product Title
3M™ First Base™ + Bump Cap